missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Methoxy-3-nitropyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
50 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Quantity | 10 g |
Formula Weight | 154.12 |
Percent Purity | 97% |
Chemical Name or Material | 2-Methoxy-3-nitropyridine |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'