Få mere at vide
2-Methoxy 17beta-Estradiol, TRC
Artikelnummer. 30137375
Skift visning
Mængde:
50 mg
100 mg
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137375 100 mg
30133271 5 g
17961065 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137375 Leverandør Toronto Research Chemicals Leverandørnr. TRCM262625100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 362-07-2
Molekylær formel C19 H26 O3
Molekylvægt (g/mol) 302.41
Synonym (17β)-2-Methoxyestra-1,3,5(10)-triene-3,17-diol, Estra-1,3,5(10)-triene-3,17β-diol, 2-methoxy- (7CI,8CI), Estradiol, 2-methoxy- (6CI), (17β)-2-Methoxyestra-1,3,5(10)-triene-3,17-diol, 2-Hydroxyestradiol 2-methyl ether, 2-Methoxy-E2, 2-Methoxyestra-1,3,5(10)-triene-3,17β-diol, 2-Methoxyestradiol, 2ME2, NSC 659853, Panzem
IUPAC navn (8R,9S,13S,14S,17S)-2-methoxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-3,17-diol
SMIL COc1cc2[C@H]3CC[C@]4(C)[C@@H](O)CC[C@H]4[C@@H]3CCc2cc1O
Tekniske data
Kemisk navn eller materiale 2-Methoxyestradiol
Mængde 100 mg
InChI formel InChI=1S/C19H26O3/c1-19-8-7-12-13(15(19)5-6-18(19)21)4-3-11-9-16(20)17(22-2)10-14(11)12/h9-10,12-13,15,18,20-21H,3-8H2,1-2H3/t12-,13+,15-,18-,19-/m0/s1
Formel vægt 302.1882