missing translation for 'onlineSavingsMsg'
Learn More

2-Mercaptopyridine-N-oxide, 99%, Thermo Scientific Chemicals

Catalog Number p-3778943
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
1121-31-9
C5H5NOS
127.16
MFCD00006196
YBBJKCMMCRQZMA-UHFFFAOYSA-N
pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide
1570
CHEBI:36578
1-hydroxypyridine-2-thione
C1=CC(=S)N(C=C1)O
This item is not returnable. View return policy
1121-31-9
C5H5NOS
127.16
MFCD00006196
YBBJKCMMCRQZMA-UHFFFAOYSA-N
pyrithione, 2-mercaptopyridine n-oxide, 2-mercaptopyridine-n-oxide, 1-hydroxy-2-pyridinethione, omadine, 2-pyridinethiol, 1-oxide, 1-hydroxyl-1h-pyridine-2-thione, n-hydroxypyridine-2-thione, 1-hydroxy-2 1h-pyridinethione, 2-pyridinethiol 1-oxide
1570
CHEBI:36578
1-hydroxypyridine-2-thione
C1=CC(=S)N(C=C1)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 70°C to 73°C
Color Beige to White or Gray-Green or Green-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Iodometry)
Packaging Glass bottle
Quantity 1 g
Beilstein 21, V,7, 150
Fieser 12,417; 14,268; 17,292
Merck Index 14, 7994
Formula Weight 127.16
Percent Purity 99%
Physical Form Powder, Crystals or Chunks
Chemical Name or Material 2-Mercaptopyridine-N-oxide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Mercaptopyridine-N-oxide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'