Få mere at vide
2-Mercapto-4,5,6,7-d4-benzimidazole, TRC
Artikelnummer. 30472868
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30472868 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30472868 Leverandør Toronto Research Chemicals Leverandørnr. TRCM2516125MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Mercapto-4,5,6,7-d4-benzimidazole
CAS 931581-17-8
Renhedsgrad noter HPLC
Molekylær formel C7 2H4 H2 N2 S
Mængde 5 mg
Synonym 2H-Benzimidazole-2-thione-4,5,6,7-d4, 1,3-dihydro- (ACI), 1,3-Dihydro-2H-benzimidazole-2-thione-4,5,6,7-d4 (ACI), 2-Mercaptobenzimidazole-4,5,6,7-d4, 1H-Benzimidazole-2-thiol-4,5,6,7-D4, 2-Mercapto-4,5,6,7-d4-benzimidazole, 2-Mercaptobenzimidazole-4,5,6,7-D4
InChI formel InChI=1S/C7H6N2S/c10-7-8-5-3-1-2-4-6(5)9-7/h1-4H,(H2,8,9,10)/i1D,2D,3D,4D
SMIL [2H]c1c([2H])c([2H])c2[nH]c(S)nc2c1[2H]
IUPAC navn 4,5,6,7-tetradeuterio-1H-benzimidazole-2-thiol
Molekylvægt (g/mol) 154.23
Formel vægt 154.0503
Procent renhed >95