Få mere at vide
2-Linoleoyl-rac-glycerol (>80%), TRC
Artikelnummer. 30132633
Skift visning
Mængde:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30132633 25 mg
30008466 50 mg
30007250 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30132633 Leverandør Toronto Research Chemicals Leverandørnr. TRCL46796025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 3443-82-1
Molekylær formel C21 H38 O4
Molekylvægt (g/mol) 354.52
Synonym (9Z,12Z)-9,12-Octadecadienoic Acid 2-Hydroxy-1-(hydroxymethyl)ethyl Ester, 2-Linoleoylglycerol, 2-Monolinolein, 2-Monolinoleoylglycerol, β-Monolinolein
IUPAC navn 1,3-dihydroxypropan-2-yl (9Z,12Z)-octadeca-9,12-dienoate
SMIL CCCCC\C=C/C\C=C/CCCCCCCC(=O)OC(CO)CO
Tekniske data
Kemisk navn eller materiale 2-Linoleoyl Glycerol
Mængde 25 mg
InChI formel InChI=1S/C21H38O4/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-21(24)25-20(18-22)19-23/h6-7,9-10,20,22-23H,2-5,8,11-19H2,1H3/b7-6-,10-9-
Formel vægt 354.277