missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Isothiocyanatoacetamide, TRC

Artikelnummer. 30341938
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30341938 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30341938 Leverandør Toronto Research Chemicals Leverandørnr. TRCB452430100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Isothiocyanatoacetamide
CAS 145518-36-1
Molekylær formel C3H4N2OS
Mængde 100 mg
InChI formel InChI=1S/C3H4N2OS/c4-3(6)1-5-2-7/h1H2,(H2,4,6)
SMIL NC(=O)CN=C=S
IUPAC navn 2-isothiocyanatoacetamide
Molekylvægt (g/mol) 116.14
Formel vægt 116.0044
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.