Få mere at vide
2-Isopropyl-6-methyl-4-pyrimidone, TRC
Artikelnummer. 30388857
Skift visning
Mængde:
500 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30388857 500 mg
30402996 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30388857 Leverandør Toronto Research Chemicals Leverandørnr. TRCI872720500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 2814-20-2
Molekylær formel C8 H12 N2 O
Molekylvægt (g/mol) 152.19
Synonym 4(3H)-Pyrimidinone, 6-methyl-2-(1-methylethyl)- (ACI), 4(1H)-Pyrimidinone, 2-isopropyl-6-methyl- (8CI), 4(1H)-Pyrimidinone, 6-methyl-2-(1-methylethyl)- (9CI), 4-Pyrimidinol, 2-isopropyl-6-methyl- (6CI, 7CI), 6-Methyl-2-(1-methylethyl)-4(3H)-pyrimidinone (ACI), 2-Isopropyl-4-hydroxy-6-methylpyrimidine, 2-Isopropyl-4-methyl-6-hydroxypyrimidine, 2-Isopropyl-4-methyl-6-pyrimidinone, 2-Isopropyl-4-methylpyrimidin-6-ol, 2-Isopropyl-6-methyl-4(3H)-pyrimidone, 2-Isopropyl-6-methyl-4-hydroxypyrimidine, 2-Isopropyl-6-methyl-4-pyrimidinol, 2-Isopropyl-6-methyl-4-pyrimidone, 4-Hydroxy-2-isopropyl-6-methylpyrimidine, G 27550, IMHP, Pyrimidinol
IUPAC navn 6-methyl-2-propan-2-ylpyrimidin-4-ol
SMIL CC(C)c1nc(C)cc(O)n1
Tekniske data
Kemisk navn eller materiale 2-Isopropyl-6-methyl-4-pyrimidinol
Mængde 500 mg
InChI formel InChI=1S/C8H12N2O/c1-5(2)8-9-6(3)4-7(11)10-8/h4-5H,1-3H3,(H,9,10,11)
Formel vægt 152.095