Få mere at vide
2-Isopropyl-3-methoxy-d3-pyrazine, CDN
Artikelnummer. 30320076
Skift visning
Mængde:
0.01 g
Artikelnummer. Mængde
30320076 0.01 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320076 Leverandør CDN Leverandørnr. CDND69050.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-(Methoxy-d3)-3-(1-methylethyl)pyrazine
CAS 588732-60-9
Molekylær formel C8 2H3 H9 N2 O
Mængde 0.01 g
InChI formel InChI=1 S/C8H12N2O/c1-6(2)7-8(11-3)10-5-4-9-7/h4-6 H,1-3H3/i3D3
SMIL [2 H]C([2 H])([2 H])Oc1nccnc1C(C)C
IUPAC navn 2-propan-2-yl-3-(trideuteriomethoxy)pyrazine
Molekylvægt (g/mol) 155.212
Formel vægt 155.114 g/mol
Procent renhed 99 atom % D, min 97% Chemical Purity