missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Isopropoxyethanol, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
1000 mL
2500 mL
Description
2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.
Solubility
It is soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
2-Isopropoxyethanol,is used as a multipurpose intermediate, Application intermediate and also used as a chemical in organic synthesis. They are used as a solvents for many resins, oils, waxes, fats and dyestuff.
Solubility
It is soluble in water.
Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
Specifications
Melting Point | -60°C |
Density | 0.903 |
Boiling Point | 143°C |
Flash Point | 45°C (113°F) |
Linear Formula | (CH3)2CHO(CH2)2OH |
Refractive Index | 1.41 |
Quantity | 100 mL |
UN Number | UN1992 |
Beilstein | 1732184 |
Solubility Information | It is soluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'