Få mere at vide
(2-Isocyanoethyl)-benzene, TRC
Artikelnummer. 17954565
Skift visning
Mængde:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17954565 1 g
17931065 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17954565 Leverandør Toronto Research Chemicals Leverandørnr. TRCI8090001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 59795-89-0
Molekylær formel C9 H9 N
Molekylvægt (g/mol) 131.1745
Synonym Benzene, (2-isocyanoethyl)-, 1-Isocyano-2-phenylethane, 2-Phenylethyl isocyanide, Phenethyl isocyanide
IUPAC navn 2-isocyanoethylbenzene
SMIL [C-]#[N+]CCc1ccccc1
Tekniske data
Kemisk navn eller materiale (2-Isocyanoethyl)-benzene
Mængde 1 g
InChI formel InChI=1S/C9H9N/c1-10-8-7-9-5-3-2-4-6-9/h2-6H,7-8H2
Formel vægt 131.0735