Få mere at vide
2-Isobutyl-3-methoxy-d3-pyrazine, CDN
Artikelnummer. 30309225
Skift visning
Mængde:
0.025 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309225 0.025 g
30322060 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30309225 Leverandør CDN Leverandørnr. CDND59750.025G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Isobutyl-3-methoxypyrazine-D3 (methoxy-D3)
CAS 588732-63-2
Molekylær formel C9 D3 H11 N2 O
Mængde 0.025 g
Synonym Pyrazine, 2-(methoxy-d3)-3-(2-methylpropyl)- (9 CI), 2-Isobutyl-3-methoxy-d3-pyrazine, 2-(Methoxy-d3)-3-(2-methylpropyl)pyrazine, 2-Isobutyl-3-methoxypyrazine-D3 (methoxy-D3)
InChI formel InChI=1 S/C9H14N2O/c1-7(2)6-8-9(12-3)11-5-4-10-8/h4-5,7 H,6H2,1-3H3/i3D3
SMIL [2 H]C([2 H])([2 H])Oc1nccnc1CC(C)C
IUPAC navn 2-(2-methylpropyl)-3-(trideuteriomethoxy)pyrazine
Molekylvægt (g/mol) 169.2388
Formel vægt 169.1294 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity