Få mere at vide
2-iso-Propyl-d7-5-methyl-d3-phenol-3,4,6-d3, TRC
Artikelnummer. 30449143
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30449143 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30449143 Leverandør Toronto Research Chemicals Leverandørnr. TRCT4130015MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thymol-d13
CAS 1219798-93-2
Molekylær formel C10 2H13 H O
Mængde 5 mg
Synonym Thymol D13 (1-methylethyl D7)
InChI formel InChI=1S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11H,1-3H3/i1D3,2D3,3D3,4D,5D,6D,7D
SMIL [2H]c1c([2H])c(c(O)c([2H])c1C([2H])([2H])[2H])C([2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2,4,5-trideuterio-6-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-3-(trideuteriomethyl)phenol
Molekylvægt (g/mol) 163.3
Formel vægt 163.1861