Få mere at vide
2-iso-Propyl-d7-5-methyl-d3-phenol-3,4,6-d3, CDN
Artikelnummer. 30327681
Skift visning
Mængde:
0.1 g
0.25 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30327681 0.1 g 0.1g
30321348 0.25 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327681 Leverandør CDN Leverandørnr. CDND67170.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Thymol-d13
CAS 1219798-93-2
Molekylær formel C10 2H13 H O
Mængde 0.1 g
Synonym Thymol D13 (1-methylethyl D7)
InChI formel InChI=1 S/C10H14O/c1-7(2)9-5-4-8(3)6-10(9)11/h4-7,11 H,1-3H3/i1D3,2D3,3D3,4 D,5 D,6 D,7 D
SMIL [2 H]c1c([2 H])c(c(O)c([2 H])c1C([2 H])([2 H])[2 H])C([2 H])(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2,4,5-trideuterio-6-(1,1,1,2,3,3,3-heptadeuteriopropan-2-yl)-3-(trideuteriomethyl)phenol
Molekylvægt (g/mol) 163.3
Formel vægt 163.1861 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity