missing translation for 'onlineSavingsMsg'
Learn More

2-Iodopyrazine, 95%, Thermo Scientific Chemicals

Catalog Number p-7028834
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
32111-21-0
C4H3IN2
205.986
MFCD01319019
OYWPFIUVDKHHGQ-UHFFFAOYSA-N
iodopyrazine, 2-iodo-pyrazine, 2-iodo-1,3-diazine, iodo-pyrazine, 2-iodo pyrazine, pyrazine, 2-iodo, acmc-1ad2s
642841
2-iodopyrazine
C1=CN=C(C=N1)I
This item is not returnable. View return policy
32111-21-0
C4H3IN2
205.986
MFCD01319019
OYWPFIUVDKHHGQ-UHFFFAOYSA-N
iodopyrazine, 2-iodo-pyrazine, 2-iodo-1,3-diazine, iodo-pyrazine, 2-iodo pyrazine, pyrazine, 2-iodo, acmc-1ad2s
642841
2-iodopyrazine
C1=CN=C(C=N1)I

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 2.086
Boiling Point 109°C to 110°C (34 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.655
Quantity 1 g
Sensitivity Light sensitive
Formula Weight 205.98
Percent Purity 95%
Chemical Name or Material 2-Iodopyrazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'