Få mere at vide
2-Iodoethanol-1,1,2,2-d4, CDN
Artikelnummer. 30324591
Skift visning
Mængde:
0.05 g
0.1 g
Pakningsstørrelse:
0.1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30324591 0.1 g 0.1g
30321291 0.05 g -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324591 Leverandør CDN Leverandørnr. CDND82390.1G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Iodoethanol D4
CAS 284474-46-0
Molekylær formel C2 D4 H I O
Mængde 0.1 g
Synonym 2-Iodoethanol D4, Ethan-1,1,2,2-d4-ol, 2-iodo-, 2-Iodoethanol-1,1,2,2-d4
InChI formel InChI=1 S/C2H5IO/c3-1-2-4/h4H,1-2H2/i1D2,2D2
SMIL [2 H]C([2 H])(O)C([2 H])([2 H])I
IUPAC navn 1,1,2,2-tetradeuterio-2-iodoethanol
Molekylvægt (g/mol) 175.9896
Formel vægt 175.9636 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity