Learn More
2-Iodobutane, 99%, stab. with copper, Thermo Scientific Chemicals
Catalog Number p-7025234
Click to view available options
Quantity:
50 g
250 g
500 g
513-48-4
C4H9I
184.02
MFCD00001072
IQRUSQUYPCHEKN-UHFFFAOYNA-N
sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide
10559
2-iodobutane
CCC(C)I
513-48-4
C4H9I
184.02
MFCD00001072
IQRUSQUYPCHEKN-UHFFFAOYNA-N
sec-butyl iodide, butane, 2-iodo, 2-butyl iodide, sec-iodobutane, 2-jodbutan, 2-jodbutan czech, 2-iodanylbutane, 2-iodobutan, 2-iodo-butane, sec.-butyl iodide
10559
2-iodobutane
CCC(C)I
It is used as a solvent organic synthesis and as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used as a solvent organic synthesis and as pharmaceutical intermediate.
Solubility
Insoluble in water. Soluble in alcohol and diethyl ether.
Notes
Store at room temperature. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point -104°C
Density 1.597
Boiling Point 119°C to 120°C
Flash Point 21°C (70°F)
Odor Ether-like
Linear Formula CH3CH2CHICH3
Refractive Index 1.4991
Quantity 50 g
UN Number UN2390
Beilstein 1718777
Sensitivity Light sensitive
Merck Index 14,1574
Solubility Information Insoluble in water. Soluble in alcohol and diethyl ether.
Formula Weight 184.02
Percent Purity 99%
Chemical Name or Material 2-Iodobutane, stabilized with copper