missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Iodo-6-methoxy-4-(2-thiazolidinyl)phenol, 97%

Brand:  Alfa Aesar™ H62275.06

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15408828

  • 6005.85 DKK / 5g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

2-Iodo-6-methoxy-4-(2-thiazolidinyl)phenol
337.18
MFCD26960673
2-(4-Hydroxy-3-iodo-5-methoxyphenyl)thiazolidine

Specifications

2-Iodo-6-methoxy-4-(2-thiazolidinyl)phenol
97%
MFCD26960673
2-(4-Hydroxy-3-iodo-5-methoxyphenyl)thiazolidine
Solid
337.18
5g
Light Sensitive
Videos
SDS