missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Iodo-4-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
The 10-chloro-8-fluoro-10, 11-dihydro-2-trifluoromethyldibenzothiepin used as the starting material can be prepared starting from 2-iodo--4-trifluoromethyl-benzoic acid and 4-fluoro-thiophenol.
Solubility
Slightly soluble.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
The 10-chloro-8-fluoro-10, 11-dihydro-2-trifluoromethyldibenzothiepin used as the starting material can be prepared starting from 2-iodo--4-trifluoromethyl-benzoic acid and 4-fluoro-thiophenol.
Solubility
Slightly soluble.
Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
Specifications
Quantity | 1 g |
Sensitivity | Light sensitive |
Solubility Information | Slightly soluble. |
Formula Weight | 316.02 |
Percent Purity | 98% |
Chemical Name or Material | 2-Iodo-4-(trifluoromethyl)benzoic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'