missing translation for 'onlineSavingsMsg'
Learn More

2-Iodo-1-methylimidazole, 97%, Thermo Scientific Chemicals

Catalog Number p-7034195
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
37067-95-1
C4H5IN2
208.002
MFCD00661606
BKRACZNEJSCZML-UHFFFAOYSA-N
2-iodo-1-methyl-1h-imidazole, 1h-imidazole, 2-iodo-1-methyl, pubchem16310, acmc-1ajih, 2-iodo-1-methyl-imidazole, 2-iodanyl-1-methyl-imidazole, 1-methyl-2-iodo-1h-imidazole, 2-iodo-n-methylimidazole, 1-methyl-2-iodoimidazole, 1h-imidazole,2-iodo-1-methyl
335835
2-iodo-1-methylimidazole
CN1C=CN=C1I
This item is not returnable. View return policy
37067-95-1
C4H5IN2
208.002
MFCD00661606
BKRACZNEJSCZML-UHFFFAOYSA-N
2-iodo-1-methyl-1h-imidazole, 1h-imidazole, 2-iodo-1-methyl, pubchem16310, acmc-1ajih, 2-iodo-1-methyl-imidazole, 2-iodanyl-1-methyl-imidazole, 1-methyl-2-iodo-1h-imidazole, 2-iodo-n-methylimidazole, 1-methyl-2-iodoimidazole, 1h-imidazole,2-iodo-1-methyl
335835
2-iodo-1-methylimidazole
CN1C=CN=C1I

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 79°C to 82°C
Quantity 250 mg
Sensitivity Light sensitive
Formula Weight 208
Percent Purity 97%
Chemical Name or Material 2-Iodo-1-methylimidazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'