missing translation for 'onlineSavingsMsg'
Learn More

2-Imidazolidinethione, 98%, Thermo Scientific Chemicals

Catalog Number p-147950
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
96-45-7
C3H6N2S
102.16
MFCD00005276
PDQAZBWRQCGBEV-UHFFFAOYSA-N
2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline
2723650
CHEBI:34750
imidazolidine-2-thione
S=C1NCCN1
This item is not returnable. View return policy
96-45-7
C3H6N2S
102.16
MFCD00005276
PDQAZBWRQCGBEV-UHFFFAOYSA-N
2-imidazolidinethione, ethylenethiourea, ethylene thiourea, 2-mercaptoimidazoline, imidazolidinethione, n,n'-ethylenethiourea, mercazin i, warecure c, rhenogran etu, mercaptoimidazoline
2723650
CHEBI:34750
imidazolidine-2-thione
S=C1NCCN1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 197.0°C to 201.0°C
Color White
Flash Point 252°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 250 g
Beilstein 24, 4
Merck Index 15, 3858
Solubility Information Solubility in water: 19g/L (20°C). Other solubilities: 440g/L water (90°C), moderately soluble in methanol, ethanol, ethylene, glycol and pyridine, insoluble in acetone, ether, chloroform, benzene, and ligroin
Formula Weight 102.15
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Imidazolidinethione, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Imidazolidinethione
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
Hazard Statement
  • H360D-May damage the unborn child.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'