Få mere at vide
(2-Hydroxypropyl)-gamma-cyclodextrin (Technical Grade), TRC
Artikelnummer. 30009698
Skift visning
Mængde:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30009698 1 g
30009499 5 g
30007785 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30009698 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9521201G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 128446-34-4
Molekylær formel C7H6FNO2
Molekylvægt (g/mol) 155.13
Synonym 2-hydroxypropyl Ethers γ-Cyclodextrin
SMIL CC1=CC(=C(C=C1)[N+](=O)[O-])F
Tekniske data
Kemisk navn eller materiale (2-Hydroxypropyl)-gamma-cyclodextrin
Mængde 1 g
InChI formel InChI=1S/C63H110O45/c1-19(67)9-88-14-27-51-35(75)43(83)59(96-27)102-49-25(7-65)93-56(40(80)32(49)72)101-48-24(6-64)94-57(41(81)33(48)73)105-52-28(15-89-10-20(2)68)98-61(45(85)37(52)77)107-54-30(17-91-12-22(4)70)100-63(47(87)39(54)79)108-55-31(18-92-13-23(5)71)99-62(46(86)38(55)78)106-53-29(16-90-11-21(3)69)97-60(44(84)36(53)76)103-50-26(8-66)95-58(104-51)42(82)34(50)74/h19-87H,6-18H2,1-5H3/t19?,20?,21?,22?,23?,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-,43-,44-,45-,46-,47-,48-,49-,50-,51-,52-,53-,54-,55-,56-,57-,58-,59-,60-,61-,62-,63-/m1/s1
Formel vægt 1586.6319