Få mere at vide
2-Hydroxymethyl-5-(methoxycarbonyl)-6-methyl-4-(2-nitrophenyl)pyridine-3-carboxylic Acid Lactone (Hydroxy Dehydro Nifedipine Lactone), Mikromol™
Beskrivelse
2-Hydroxymethyl-5-(methoxycarbonyl)-6-methyl-4-(2-nitrophenyl)pyridine-3-carboxylic Acid Lactone (Hydroxy Dehydro Nifedipine Lactone)
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Hydroxymethyl-5-(methoxycarbonyl)-6-methyl-4-(2-nitrophenyl)pyridine-3-carboxylic Acid Lactone |
| CAS | 34785-00-7 |
| Analyt- eller komponentnavne | 2-Hydroxymethyl-5-(methoxycarbonyl)-6-methyl-4-(2-nitrophenyl)pyridine-3-carboxylic Acid Lactone |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C16 H12 N2 O6 |
| Certificeringer/compliance | ISO 9001 |
| Synonym | Furo[3,4-b]pyridine-3-carboxylic acid, 5,7-dihydro-2-methyl-4-(2-nitrophenyl)-5-oxo-, methyl ester, : 4-(2-Nitrophenyl)-2-(hydroxymethyl)-5-(methoxycarbonyl)-6-methylpyridine-3-carboxylic acid-γ-lactone, BAY-h 2228, 2-Hydroxymethyl-5-(methoxycarbonyl)-6-methyl-4-(2-nitrophenyl)pyridine-3-carboxylic Acid Lactone |
| InChI formel | InChI=1S/C16H12N2O6/c1-8-12(15(19)23-2)13(14-10(17-8)7-24-16(14)20)9-5-3-4-6-11(9)18(21)22/h3-6H,7H2,1-2H3 |
| SMIL | COC(=O)c1c(C)nc2COC(=O)c2c1c3ccccc3[N+](=O)[O-] |
| IUPAC navn | methyl 2-methyl-4-(2-nitrophenyl)-5-oxo-7H-furo[3,4-b]pyridine-3-carboxylate |
| Vis mere |