missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxymethyl-1,4-benzodioxane, 98%, Thermo Scientific Chemicals

Catalog Number p-7021717
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
3663-82-9
C9H10O3
166.176
MFCD00006822
GWQOQQVKVOOHTI-UHFFFAOYSA-N
2-hydroxymethyl-1,4-benzodioxane, 2,3-dihydro-benzo 1,4 dioxin-2-yl-methanol, 1,4-benzodioxan-2-methanol, 2,3-dihydro-1,4-benzodioxin-2-ylmethanol, 2,3-dihydrobenzo b 1,4 dioxin-2-yl methanol, 1,4-benzodioxane-2-methanol, 2-hydroxymethyl-1,4-benzodioxan, 1,4-benzodioxin-2-methanol, 2,3-dihydro
19314
2,3-dihydro-1,4-benzodioxin-3-ylmethanol
C1C(OC2=CC=CC=C2O1)CO
This item is not returnable. View return policy
3663-82-9
C9H10O3
166.176
MFCD00006822
GWQOQQVKVOOHTI-UHFFFAOYSA-N
2-hydroxymethyl-1,4-benzodioxane, 2,3-dihydro-benzo 1,4 dioxin-2-yl-methanol, 1,4-benzodioxan-2-methanol, 2,3-dihydro-1,4-benzodioxin-2-ylmethanol, 2,3-dihydrobenzo b 1,4 dioxin-2-yl methanol, 1,4-benzodioxane-2-methanol, 2-hydroxymethyl-1,4-benzodioxan, 1,4-benzodioxin-2-methanol, 2,3-dihydro
19314
2,3-dihydro-1,4-benzodioxin-3-ylmethanol
C1C(OC2=CC=CC=C2O1)CO

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 87°C to 91°C
Quantity 5 g
Beilstein 83736
Formula Weight 166.18
Percent Purity 98%
Chemical Name or Material 2-Hydroxymethyl-1,4-benzodioxane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'