Få mere at vide
2-Hydroxyatrazine-d5 (ethyl-d5), CDN
Artikelnummer. 30327351
Skift visning
Mængde:
0.005 g
0.01 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327351 0.005 g
30323421 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30327351 Leverandør CDN Leverandørnr. CDND73180.005G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Atrazine-2-hydroxyD5
CAS 1276197-25-1
Molekylær formel C8 2H5 H10 N5 O
Mængde 0.005 g
Synonym Atrazine-2-hydroxy D5, Atrazine-2-hydroxy D5 (ethyl D5), 4-(Isopropylamino)-6-(1,1,2,2,2-pentadeuterioethylamino)-1,3,5-triazin-2-ol
InChI formel InChI=1 S/C8H15N5O/c1-4-9-6-11-7(10-5(2)3)13-8(14)12-6/h5H,4H2,1-3H3,(H3,9,10,11,12,13,14)/i1D3,4D2
SMIL [2 H]C([2 H])([2 H])C([2 H])([2 H])Nc1nc(O)nc(NC(C)C)n1
IUPAC navn 4-(1,1,2,2,2-pentadeuterioethylamino)-6-(propan-2-ylamino)-1,3,5-triazin-2-ol
Molekylvægt (g/mol) 202.27
Formel vægt 202.159 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity