Få mere at vide
2-Hydroxy Trimipramine-d3, TRC
Artikelnummer. 30405792
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30405792 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30405792 Leverandør Toronto Research Chemicals Leverandørnr. TRCH97350210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Hydroxy Trimipramine-d3
CAS 1189647-53-7
Molekylær formel C20 D3 H23 N2 O
Mængde 10 mg
InChI formel InChI=1S/C20H26N2O/c1-15(13-21(2)3)14-22-19-7-5-4-6-16(19)8-9-17-12-18(23)10-11-20(17)22/h4-7,10-12,15,23H,8-9,13-14H2,1-3H3/i2D3
SMIL [2H]C([2H])([2H])N(C)CC(C)CN1c2ccccc2CCc3cc(O)ccc13
IUPAC navn 11-[2-methyl-3-[methyl(trideuteriomethyl)amino]propyl]-5,6-dihydrobenzo[b][1]benzazepin-3-ol
Molekylvægt (g/mol) 313.45
Formel vægt 313.2233