Få mere at vide
2-Hydroxy-5-Amino-N-(2-carboxy-4-aminophenyl)benzoic Acid, TRC
Artikelnummer. 30407834
Skift visning
Mængde:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30407834 10 mg
30399543 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30407834 Leverandør Toronto Research Chemicals Leverandørnr. TRCH76800010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1797983-23-3
Molekylær formel C14 H12 N2 O5
Molekylvægt (g/mol) 288.26
Synonym 5-Amino-2-((3-carboxy-4-hydroxyphenyl)amino)benzoic Acid, N-(2-Carboxy-4-aminophenyl)-5-aminosalicylic Acid, 2-Hydroxy-5-[(2-carboxy-4-aminophenyl)amino]benzoic Acid
IUPAC navn 5-amino-2-(3-carboxy-4-hydroxyanilino)benzoic acid
SMIL Nc1ccc(Nc2ccc(O)c(c2)C(=O)O)c(c1)C(=O)O
Tekniske data
Kemisk navn eller materiale 2-Hydroxy-5-[(2-carboxy-4-aminophenyl)amino]benzoic acid
Mængde 10 mg
InChI formel InChI=1S/C14H12N2O5/c15-7-1-3-11(9(5-7)13(18)19)16-8-2-4-12(17)10(6-8)14(20)21/h1-6,16-17H,15H2,(H,18,19)(H,20,21)
Formel vægt 288.0746