missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoic Acid Hydrochloride, Mikromol™
Beskrivelse
2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoic Acid Hydrochloride
Tekniske data
Tekniske data
| Kemisk navn eller materiale | 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoic Acid Hydrochloride |
| CAS | 2726492-68-6 |
| Analyt- eller komponentnavne | 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoic Acid Hydrochloride |
| Forsendelsestilstand | Room Temperature |
| Molekylær formel | C19 H23 N O4 . Cl H |
| Certificeringer/compliance | ISO 9001 |
| Synonym | 2-Hydroxy-5-[1-hydroxy-2-[(1-methyl-3-phenylpropyl)amino]ethyl]benzoic Acid Hydrochloride, Labetalol Hydrochloride Imp. A (EP) as Hydrochloride, Labetalol Hydrochloride Impurity A as Hydrochloride, Labetalol Impurity A as Hydrochloride |
| InChI formel | InChI=1S/C19H23NO4.ClH/c1-13(7-8-14-5-3-2-4-6-14)20-12-18(22)15-9-10-17(21)16(11-15)19(23)24;/h2-6,9-11,13,18,20-22H,7-8,12H2,1H3,(H,23,24);1H |
| SMIL | Cl.CC(CCc1ccccc1)NCC(O)c2ccc(O)c(c2)C(=O)O |
| IUPAC navn | 2-hydroxy-5-[1-hydroxy-2-(4-phenylbutan-2-ylamino)ethyl]benzoic acid;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?