Learn More
Alfa Aesar™ 2-Hydroxy-4-(trifluoromethyl)pyrimidine, 99%
Brand: Alfa Aesar™ H60151.06
Additional Details : CAS Number : 104048-92-2 Weight : 0.00500kg
Quantity | 5g |
---|
Chemical Identifiers
104048-92-2 | |
164.087 | |
WCEBFEVZTGLOHC-UHFFFAOYSA-N | |
1240050 | |
C1=C(NC(=O)N=C1)C(F)(F)F |
C5H3F3N2O | |
MFCD00192529 | |
2-hydroxy-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ol, 4-trifluoromethyl pyrimidin-2 1h-one, 2-hydroxy-4-trifluoromethylpyrimidine, 4-trifluoromethyl-2-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-2-one, 2 1h-pyrimidinone, 4-trifluoromethyl, pubchem7201, enamine_002404, acmc-2098am | |
6-(trifluoromethyl)-1H-pyrimidin-2-one |
Specifications
2-Hydroxy-4-(trifluoromethyl)pyrimidine | |
5g | |
C5H3F3N2O | |
2-hydroxy-4-trifluoromethyl pyrimidine, 4-trifluoromethyl pyrimidin-2-ol, 4-trifluoromethyl pyrimidin-2 1h-one, 2-hydroxy-4-trifluoromethylpyrimidine, 4-trifluoromethyl-2-pyrimidinol, 6-trifluoromethyl-1h-pyrimidin-2-one, 2 1h-pyrimidinone, 4-trifluoromethyl, pubchem7201, enamine_002404, acmc-2098am | |
C1=C(NC(=O)N=C1)C(F)(F)F | |
164.087 | |
164.09 |
223°C to 227°C | |
104048-92-2 | |
MFCD00192529 | |
WCEBFEVZTGLOHC-UHFFFAOYSA-N | |
6-(trifluoromethyl)-1H-pyrimidin-2-one | |
1240050 | |
99% |