Få mere at vide
2-Hydroxy-4-quinolinecarboxylic Acid Methyl Ester, Mikromol™
Artikelnummer. 30049232
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30049232 25 mg
30049438 100 mg
30457808 4 x 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30049232 Leverandør Mikromol Leverandørnr. MM0604.070025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2-Hydroxy-4-quinolinecarboxylic Acid Methyl Ester
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 2-Hydroxy-4-quinolinecarboxylic Acid Methyl Ester
CAS 84906-82-1
Analyt- eller komponentnavne 2-Hydroxy-4-quinolinecarboxylic Acid Methyl Ester
Forsendelsestilstand Room Temperature
Molekylær formel C11 H9 N O3
Certificeringer/compliance ISO 17025
Synonym 2-Hydroxy-4-quinolinecarboxylic Acid Methyl Ester, Cinchocaine Hydrochloride Imp. B (EP) as Methyl Ester
InChI formel InChI=1S/C11H9NO3/c1-15-11(14)8-6-10(13)12-9-5-3-2-4-7(8)9/h2-6H,1H3,(H,12,13)
SMIL COC(=O)c1cc(O)nc2ccccc12
IUPAC navn methyl 2-hydroxyquinoline-4-carboxylate
Molekylvægt (g/mol) 203.19
Mængde 25 mg
Formel vægt 203.0582 g/mol