missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Hydroxy-4-phenylbutyric Acid Ethyl Ester-d5, TRC

Artikelnummer. 30407438
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30407438 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30407438 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9491025MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale 2-Hydroxy-4-phenylbutyric Acid Ethyl Ester-d5
CAS 1189892-05-4
Molekylær formel C12H11D5O3
Mængde 5 mg
Synonym alpha-Hydroxybenzenebutanoic Acid-d5 Ethyl Ester, Ethyl 2-Hydroxy-4-phenylbutanoate-d5, Ethyl 2-Hydroxy-4-phenylbutyrate-d5, Ethyl 4-Phenyl-2-hydroxybutyrate-d5, Ethyl alpha-Hydroxybenzenebutanoate-d5
InChI formel InChI=1S/C12H16O3/c1-2-15-12(14)11(13)9-8-10-6-4-3-5-7-10/h3-7,11,13H,2,8-9H2,1H3/i3D,4D,5D,6D,7D
SMIL [2H]c1c([2H])c([2H])c(CCC(O)C(=O)OCC)c([2H])c1[2H]
IUPAC navn ethyl 2-hydroxy-4-(2,3,4,5,6-pentadeuteriophenyl)butanoate
Molekylvægt (g/mol) 213.28
Formel vægt 213.1413
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.