missing translation for 'onlineSavingsMsg'
Learn More

2-Hydroxy-4-methyl-3-nitropyridine, 98%, Thermo Scientific™

Catalog Number 15488138
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
21901-18-8
C6H6N2O3
154.13
MFCD00010689,MFCD00955770
HZCWTTHQRMHIIE-UHFFFAOYSA-N
2-hydroxy-4-methyl-3-nitropyridine, 4-methyl-3-nitropyridin-2-ol, 2-hydroxy-3-nitro-4-picoline, 2 1h-pyridinone, 4-methyl-3-nitro, 2-hydroxy-3-nitro-4-methylpyridine, 4-methyl-3-nitro-2-pyridone, 4-methyl-3-nitro-2-pyridinone, 4-methyl-3-nitro-2-pyridinol, 4-methyl-3-nitropyridin-2 1h-one, 4-methyl-3-nitro-1,2-dihydropyridin-2-one
345370
4-methyl-3-nitro-1H-pyridin-2-one
CC1=C(C(=O)NC=C1)[N+]([O-])=O
This item is not returnable. View return policy
21901-18-8
C6H6N2O3
154.13
MFCD00010689,MFCD00955770
HZCWTTHQRMHIIE-UHFFFAOYSA-N
2-hydroxy-4-methyl-3-nitropyridine, 4-methyl-3-nitropyridin-2-ol, 2-hydroxy-3-nitro-4-picoline, 2 1h-pyridinone, 4-methyl-3-nitro, 2-hydroxy-3-nitro-4-methylpyridine, 4-methyl-3-nitro-2-pyridone, 4-methyl-3-nitro-2-pyridinone, 4-methyl-3-nitro-2-pyridinol, 4-methyl-3-nitropyridin-2 1h-one, 4-methyl-3-nitro-1,2-dihydropyridin-2-one
345370
4-methyl-3-nitro-1H-pyridin-2-one
CC1=C(C(=O)NC=C1)[N+]([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 229°C to 232°C
Color Yellow
Quantity 1 g
Beilstein 139125
Formula Weight 154.12
Percent Purity 98%
Physical Form Crystalline Solid
Chemical Name or Material 2-Hydroxy-4-methyl-3-nitropyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'