Få mere at vide
2-Hydroxy-2-(o-chlorophenyl)cyclohexanone, TRC
Artikelnummer. 30135449
Skift visning
Mængde:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30135449 50 mg
30133930 100 mg
17962615 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30135449 Leverandør Toronto Research Chemicals Leverandørnr. TRCH94918050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1823362-29-3
Molekylær formel C12 H13 Cl O2
Molekylvægt (g/mol) 224.68
Synonym (2RS)-2-(2-Chlorophenyl)-2-hydroxycyclohexanone, Ketamine Hydrochloride Imp. B (EP), 2-(2-Chlorophenyl)-2-hydroxycyclohexanone (ACI), Cyclohexanone, 2-(2-chlorophenyl)-2-hydroxy- (ACI), 2-(2-Chlorophenyl)-2-hydroxycyclohexanone (ACI), 2-(2-Chlorophenyl)-2-hydroxycyclohexan-1-one
IUPAC navn 2-(2-chlorophenyl)-2-hydroxycyclohexan-1-one
SMIL OC1(CCCCC1=O)c2ccccc2Cl
Tekniske data
Kemisk navn eller materiale (2RS)-2-(2-Chlorophenyl)-2-hydroxycyclohexanone
Renhedsgrad noter HPLC
Mængde 50 mg
InChI formel InChI=1S/C12H13ClO2/c13-10-6-2-1-5-9(10)12(15)8-4-3-7-11(12)14/h1-2,5-6,15H,3-4,7-8H2
Formel vægt 224.0604
Procent renhed >95%