Få mere at vide
2-Hydroxy-17Beta-estradiol, TRC
Artikelnummer. 30008146
Skift visning
Mængde:
1 mg
2.5 mg
5 mg
Pakningsstørrelse:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
30008146 2.5 mg 2.5mg
30003741 5 mg -
30010951 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30008146 Leverandør Toronto Research Chemicals Leverandørnr. TRCH9418902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 362-05-0
Molekylær formel C18 H24 O3
Molekylvægt (g/mol) 288.38
Synonym Estra-1,3,5(10)-triene-2,3,17-triol, (17β)-, Estra-1,3,5(10)-triene-2,3,17β-triol (7CI,8CI), (17β)-Estra-1,3,5(10)-triene-2,3,17-triol, 2,3,17β-Trihydroxyestra-1,3,5(10)-triene, 2-Hydroxyestradiol, NSC 61711, 2-Hydroxy-17β-estradiol
IUPAC navn (8R,9S,13S,14S,17S)-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthrene-2,3,17-triol
SMIL C[C@]12CC[C@H]3[C@@H](CCc4cc(O)c(O)cc34)[C@@H]1CC[C@@H]2O
Tekniske data
Kemisk navn eller materiale 2-Hydroxy-17 beta-estradiol
Mængde 2.5 mg
InChI formel InChI=1S/C18H24O3/c1-18-7-6-11-12(14(18)4-5-17(18)21)3-2-10-8-15(19)16(20)9-13(10)11/h8-9,11-12,14,17,19-21H,2-7H2,1H3/t11-,12+,14-,17-,18-/m0/s1
Formel vægt 288.1725