missing translation for 'onlineSavingsMsg'
Få mere at vide

2-Hydrazino-1H-1,3-benzimidazole, TRC

Artikelnummer. 30395598
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30395598 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30395598 Leverandør Toronto Research Chemicals Leverandørnr. TRCH714553100MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemisk navn eller materiale 2-Hydrazino-1H-1,3-benzimidazole
CAS 15108-18-6
Molekylær formel C7H8N4
Mængde 100 mg
InChI formel InChI=1S/C7H8N4/c8-11-7-9-5-3-1-2-4-6(5)10-7/h1-4H,8H2,(H2,9,10,11)
SMIL NNc1nc2ccccc2[nH]1
IUPAC navn 1H-benzimidazol-2-ylhydrazine
Molekylvægt (g/mol) 148.17
Formel vægt 148.0749
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.