Learn More
2-Furoic acid, 98%
Product Code. 11440323
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11440323 100 g 100g
11450323 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11440323 Supplier Thermo Scientific Alfa Aesar Supplier No. A11454.22
 Restricted Item
På lager
Add to basket
CAS: 88-14-2 | C5H4O3 | 112.084 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88-14-2
Molecular Formula C5H4O3
Molecular Weight (g/mol) 112.084
MDL Number MFCD00003238
InChI Key SMNDYUVBFMFKNZ-UHFFFAOYSA-N
Synonym 2-furoic acid, 2-furancarboxylic acid, pyromucic acid, 2-carboxyfuran, furoic acid, furancarboxylic acid, alpha-furoic acid, alpha-furancarboxylic acid, 2-furanoic acid, kyselina 2-furoova
PubChem CID 6919
ChEBI CHEBI:30845
IUPAC Name furan-2-carboxylic acid
SMILES C1=COC(=C1)C(=O)O
Specifications
Melting Point 127°C to 132°C
Boiling Point 230°C to 232°C (sublimation)
Flash Point 137°C (278°F)
Odor Odorless
Quantity 100 g
Beilstein 110149
Merck Index 14,4307
Formula Weight 112.08
Percent Purity 98%
Chemical Name or Material 2-Furoic acid