Få mere at vide
2-Fluorobenzoic Acid-d4, TRC
Artikelnummer. 30408264
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30408264 10 mg
30406331 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30408264 Leverandør Toronto Research Chemicals Leverandørnr. TRCF58838210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 646502-89-8
Molekylær formel C7 2H4 H F O2
Molekylvægt (g/mol) 144.14
Synonym Benzoic-2,3,4,5-d4 acid, 6-fluoro- (9CI), 2-Fluorobenzoic Acid-d4, 6-Fluorobenzoic-2,3,4,5-d4 acid, 2-Fluorobenzoic-d4 Acid, 2-Fluorobenzoic Acid-D4
IUPAC navn 2,3,4,5-tetradeuterio-6-fluorobenzoic acid
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(F)c1[2H]
Tekniske data
Kemisk navn eller materiale 2-Fluorobenzoic acid ring-d4
Mængde 10 mg
InChI formel InChI=1S/C7H5FO2/c8-6-4-2-1-3-5(6)7(9)10/h1-4H,(H,9,10)/i1D,2D,3D,4D
Formel vægt 144.0525