missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluoro-N,N-dimethyl-6-nitroaniline, 97%

Brand:  Alfa Aesar™ H63011.03

Additional Details : Weight : 0.00100kg Transport : UN number : 2811 Chem class : 6.1 Pack group : III

Quantity 1g

 View more versions of this product

Product Code. 15469438

  • 564.44 DKK / 1g


Estimated Shipment date
from Supplier 15-01-2019
Add to basket

Specifications

Chemical Identifiers

174574-15-3
(2-Fluoro-6-nitrophenyl)dimethylamine

Specifications

2-Fluoro-N,N-dimethyl-6-nitroaniline
174574-15-3
(2-Fluoro-6-nitrophenyl)dimethylamine
Solid
1g
UN2811
184.17
97%
Videos
SDS