missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific™

Catalog Number 11327609
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
352535-96-7
C7H5BF4O2
207.919
MFCD03701538
KUHVVLFCTMTYGR-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl phenylboronic acid, 2-fluoro-5-trifluoromethyl phenyl boronic acid, 2-fluoro-5-trifluoromethyl benzeneboronic acid, 2-fluoro-5-trifluoromethyl boronic acid, 2-fluoro-5-trifluoromethylphenylboronic acid, boronic acid, 2-fluoro-5-trifluoromethyl phenyl, pubchem5041, acmc-1afry
2763301
[2-fluoro-5-(trifluoromethyl)phenyl]boronic acid
B(C1=C(C=CC(=C1)C(F)(F)F)F)(O)O
This item is not returnable. View return policy
352535-96-7
C7H5BF4O2
207.919
MFCD03701538
KUHVVLFCTMTYGR-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl phenylboronic acid, 2-fluoro-5-trifluoromethyl phenyl boronic acid, 2-fluoro-5-trifluoromethyl benzeneboronic acid, 2-fluoro-5-trifluoromethyl boronic acid, 2-fluoro-5-trifluoromethylphenylboronic acid, boronic acid, 2-fluoro-5-trifluoromethyl phenyl, pubchem5041, acmc-1afry
2763301
[2-fluoro-5-(trifluoromethyl)phenyl]boronic acid
B(C1=C(C=CC(=C1)C(F)(F)F)F)(O)O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 104°C to 109°C
Quantity 1 g
Formula Weight 207.92
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'