missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)aniline, 97%, Thermo Scientific Chemicals

Catalog Number p-7023721
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
535-52-4
C7H5F4N
179.118
MFCD00007653
DRKWGMXFFCPZLW-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl aniline, 3-amino-4-fluorobenzotrifluoride, alpha,alpha,alpha,6-tetrafluoro-m-toluidine, 4-fluoro-3-aminobenzotrifluoride, 2-fluoro-5-trifluoromethyl benzenamine, 2-fluoro-5-trifluoromethyl phenylamine, benzenamine, 2-fluoro-5-trifluoromethyl, .alpha.,.alpha.,.alpha.,6-tetrafluoro-m-toluidine, pubchem1538, ksc494s7l
136349
2-fluoro-5-(trifluoromethyl)aniline
C1=CC(=C(C=C1C(F)(F)F)N)F
This item is not returnable. View return policy
535-52-4
C7H5F4N
179.118
MFCD00007653
DRKWGMXFFCPZLW-UHFFFAOYSA-N
2-fluoro-5-trifluoromethyl aniline, 3-amino-4-fluorobenzotrifluoride, alpha,alpha,alpha,6-tetrafluoro-m-toluidine, 4-fluoro-3-aminobenzotrifluoride, 2-fluoro-5-trifluoromethyl benzenamine, 2-fluoro-5-trifluoromethyl phenylamine, benzenamine, 2-fluoro-5-trifluoromethyl, .alpha.,.alpha.,.alpha.,6-tetrafluoro-m-toluidine, pubchem1538, ksc494s7l
136349
2-fluoro-5-(trifluoromethyl)aniline
C1=CC(=C(C=C1C(F)(F)F)N)F

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 1.378
Boiling Point 210°C
Flash Point 70°C (158°F)
Odor Characteristic
Refractive Index 1.461
Quantity 1 g
UN Number UN2810
Beilstein 1950800
Formula Weight 179.12
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)aniline
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'