missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

Catalog Number 15428645
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1240256-82-9
C7H5BrF6S
315.071
MFCD16652495
ZACUKJFPSDLHTG-UHFFFAOYSA-N
2-fluoro-5-pentafluorosulfur benzyl bromide, 2-bromomethyl-1-fluoro-4-pentafluoro-??-sulfanyl benzene, 2-fluoro-5-pentafluorothio benzyl bromide
66523627
[3-(bromomethyl)-4-fluorophenyl]-pentafluoro-$l^{6}-sulfane
C1=CC(=C(C=C1S(F)(F)(F)(F)F)CBr)F
This item is not returnable. View return policy
1240256-82-9
C7H5BrF6S
315.071
MFCD16652495
ZACUKJFPSDLHTG-UHFFFAOYSA-N
2-fluoro-5-pentafluorosulfur benzyl bromide, 2-bromomethyl-1-fluoro-4-pentafluoro-??-sulfanyl benzene, 2-fluoro-5-pentafluorothio benzyl bromide
66523627
[3-(bromomethyl)-4-fluorophenyl]-pentafluoro-$l^{6}-sulfane
C1=CC(=C(C=C1S(F)(F)(F)(F)F)CBr)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 315.07
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(pentafluorothio)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'