missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-nitropyridine, 98%, Thermo Scientific Chemicals

Catalog Number p-7055593
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
456-24-6
C5H3FN2O2
142.089
MFCD03095059
XOZAJNLUAODXSP-UHFFFAOYSA-N
6-fluoro-3-nitropyridine, pyridine, 2-fluoro-5-nitro, 2-fluor-5-nitropyridin, 2-fluoro-5-nitro-pyridine, abbypharma ap-15-5253, pubchem1202, acmc-1arnr, 5-nitro-2-fluoropyridine, 2-fluoro-5-nitro pyridine
95264
2-fluoro-5-nitropyridine
C1=CC(=NC=C1[N+](=O)[O-])F
This item is not returnable. View return policy
456-24-6
C5H3FN2O2
142.089
MFCD03095059
XOZAJNLUAODXSP-UHFFFAOYSA-N
6-fluoro-3-nitropyridine, pyridine, 2-fluoro-5-nitro, 2-fluor-5-nitropyridin, 2-fluoro-5-nitro-pyridine, abbypharma ap-15-5253, pubchem1202, acmc-1arnr, 5-nitro-2-fluoropyridine, 2-fluoro-5-nitro pyridine
95264
2-fluoro-5-nitropyridine
C1=CC(=NC=C1[N+](=O)[O-])F

It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.

Solubility
Slightly soluble in water.

Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
TRUSTED_SUSTAINABILITY
Melting Point 17°C to 21°C
Color Colorless to Yellow
Density 1.41 g/mL
Boiling Point 86°C to 87°C (7 mmHg)(literature)
Refractive Index 1.525
Quantity 5 g
Beilstein 120351
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 142.09
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-nitropyridine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'