missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl bromide, 97%, Thermo Scientific™

Catalog Number p-7052482
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
1373921-05-1
C9H7BrF4O
287.052
MFCD22201038
RISYNKJCXPMUQS-UHFFFAOYSA-N
2-fluoro-5-methoxy-3-trifluoromethyl benzyl bromide, 1-bromomethyl-2-fluoro-5-methoxy-3-trifluoromethyl benzene
86277640
1-(bromomethyl)-2-fluoro-5-methoxy-3-(trifluoromethyl)benzene
COC1=CC(=C(C(=C1)CBr)F)C(F)(F)F
This item is not returnable. View return policy
1373921-05-1
C9H7BrF4O
287.052
MFCD22201038
RISYNKJCXPMUQS-UHFFFAOYSA-N
2-fluoro-5-methoxy-3-trifluoromethyl benzyl bromide, 1-bromomethyl-2-fluoro-5-methoxy-3-trifluoromethyl benzene
86277640
1-(bromomethyl)-2-fluoro-5-methoxy-3-(trifluoromethyl)benzene
COC1=CC(=C(C(=C1)CBr)F)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
UN Number UN3265
Formula Weight 287.05
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-methoxy-3-(trifluoromethyl)benzyl bromide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'