missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-iodobenzoic acid, 97%, Thermo Scientific Chemicals

Catalog Number p-7028767
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
124700-41-0
C7H4FIO2
266.01
MFCD03094517
QNNJHBNTHVHALE-UHFFFAOYSA-N
benzoic acid, 2-fluoro-5-iodo, 2-fluoro-5-iodobenzoicacid, 2-fluoro-5-iodo-benzoic acid, pubchem1385, acmc-1c24w, ksc174o6d, rarechem al bo 1479, buttpark 122\01-06, 5-iodo-2-fluorobenzoic acid, 2-fluoro-5-iodobenzoic acid
2778925
2-fluoro-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)F
This item is not returnable. View return policy
124700-41-0
C7H4FIO2
266.01
MFCD03094517
QNNJHBNTHVHALE-UHFFFAOYSA-N
benzoic acid, 2-fluoro-5-iodo, 2-fluoro-5-iodobenzoicacid, 2-fluoro-5-iodo-benzoic acid, pubchem1385, acmc-1c24w, ksc174o6d, rarechem al bo 1479, buttpark 122\01-06, 5-iodo-2-fluorobenzoic acid, 2-fluoro-5-iodobenzoic acid
2778925
2-fluoro-5-iodobenzoic acid
C1=CC(=C(C=C1I)C(=O)O)F

2-Fluoro-5-iodobenzoic acid is one of the precursor molecules were shown to generate ETD reagents via ESI followed by CID. Starting material in the synthesis of 3, 6-disubstituted 2-pyridinecarboxamide(GK activators).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoro-5-iodobenzoic acid is one of the precursor molecules were shown to generate ETD reagents via ESI followed by CID. Starting material in the synthesis of 3, 6-disubstituted 2-pyridinecarboxamide(GK activators).

Solubility
Insoluble in water.

Notes
Light Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, light.
TRUSTED_SUSTAINABILITY
Melting Point 162°C to 165°C
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 266.01
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-iodobenzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'