missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-hydroxybenzoic acid, 98%, Thermo Scientific™

Catalog Number 15463417
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
51446-30-1
C7H5FO3
156.11
MFCD01862084
CZNFABVUVHWEJF-UHFFFAOYSA-N
2-fluoro-5-hydroxybenozic acid, benzoic acid, 2-fluoro-5-hydroxy, 2-fluoro-5-hydroxy-benzoic acid, pubchem2260, acmc-1amqi, 3-carboxy-4-fluorophenol, 2-fluoro-5-hydroxybenzoic, ksc497o4d, 2-fluoro-5-hydroxybenozicacid, 6-fluoro-3-hydroxybenzoic acid
2774480
2-fluoro-5-hydroxybenzoic acid
OC(=O)C1=C(F)C=CC(O)=C1
This item is not returnable. View return policy
51446-30-1
C7H5FO3
156.11
MFCD01862084
CZNFABVUVHWEJF-UHFFFAOYSA-N
2-fluoro-5-hydroxybenozic acid, benzoic acid, 2-fluoro-5-hydroxy, 2-fluoro-5-hydroxy-benzoic acid, pubchem2260, acmc-1amqi, 3-carboxy-4-fluorophenol, 2-fluoro-5-hydroxybenzoic, ksc497o4d, 2-fluoro-5-hydroxybenozicacid, 6-fluoro-3-hydroxybenzoic acid
2774480
2-fluoro-5-hydroxybenzoic acid
OC(=O)C1=C(F)C=CC(O)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 204°C to 206°C
Quantity 250 mg
Formula Weight 156.11
Percent Purity 98%
Chemical Name or Material 2-Fluoro-5-hydroxybenzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'