Learn More
Alfa Aesar™ 2-Fluoro-4-(pentafluorothio)benzonitrile, 97%
Brand: Alfa Aesar™ H33862.06
Additional Details : Weight : 0.00500kg Transport : UN number : 3439 Chem class : 6.1 Pack group : III
Quantity | 5g |
---|
Chemical Identifiers
1240256-91-0 | |
247.158 | |
OVDJLPWMXSFOJG-UHFFFAOYSA-N | |
66523489 | |
C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)C#N |
C7H3F6NS | |
MFCD16652501 | |
2-fluoro-4-pentafluorosulfur benzonitrile, 2-fluoro-4-pentafluoro-??-sulfanyl benzonitrile, 2-fluoro-4-pentafluorothio benzonitrile | |
2-fluoro-4-(pentafluoro-$l^{6}-sulfanyl)benzonitrile |
Specifications
2-Fluoro-4-(pentafluorothio)benzonitrile | |
1240256-91-0 | |
MFCD16652501 | |
2-fluoro-4-pentafluorosulfur benzonitrile, 2-fluoro-4-pentafluoro-??-sulfanyl benzonitrile, 2-fluoro-4-pentafluorothio benzonitrile | |
C1=CC(=C(C=C1S(F)(F)(F)(F)F)F)C#N | |
247.158 | |
247.16 |
5g | |
C7H3F6NS | |
UN3439 | |
OVDJLPWMXSFOJG-UHFFFAOYSA-N | |
2-fluoro-4-(pentafluoro-$l^{6}-sulfanyl)benzonitrile | |
66523489 | |
97% |