missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-4-nitrotoluene, 98%, Thermo Scientific Chemicals
114.76 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.
Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
It finds its use in the synthesis of 2-fluoro-4-nitrobenzoic acid and also was used in a study of aquatic toxicity against Poecilia reticulata and Tetrahymena pyriformis.
Solubility
Insoluble in water. Solubility in methanol gives very faint turbidity.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Specifications
Melting Point | 32°C to 37°C |
Boiling Point | 56°C to 58°C (1 mmHg) |
Flash Point | 73°C (163°F) |
Quantity | 5 g |
UN Number | UN2811 |
Beilstein | 2250156 |
Solubility Information | Insoluble in water. Solubility in methanol gives very faint turbidity. |
Formula Weight | 155.13 |
Percent Purity | 98% |
Chemical Name or Material | 2-Fluoro-4-nitrotoluene |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'