missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrophenol, 98+%, Thermo Scientific Chemicals

Catalog Number 11494494
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
403-19-0
C6H4FNO3
157.1
MFCD00051970
ORPHLVJBJOCHBR-UHFFFAOYSA-N
2-fluoro-4-nitrophenol, phenol, 2-fluoro-4-nitro, 3-fluoro-4-hydroxynitrobenzene, 2-fluoro-4-nitro-phenol, pubchem4122, 4-nitro-2-fluorophenol, 2-fluoro-4-nitro phenol, acmc-1cb9t, ksc493s7t
9825
C1=CC(=C(C=C1[N+](=O)[O-])F)O
This item is not returnable. View return policy
403-19-0
C6H4FNO3
157.1
MFCD00051970
ORPHLVJBJOCHBR-UHFFFAOYSA-N
2-fluoro-4-nitrophenol, phenol, 2-fluoro-4-nitro, 3-fluoro-4-hydroxynitrobenzene, 2-fluoro-4-nitro-phenol, pubchem4122, 4-nitro-2-fluorophenol, 2-fluoro-4-nitro phenol, acmc-1cb9t, ksc493s7t
9825
C1=CC(=C(C=C1[N+](=O)[O-])F)O

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 117°C to 121°C
Quantity 1 g
Beilstein 1944995
Formula Weight 157.1
Percent Purity ≥98%
Chemical Name or Material 2-Fluoro-4-nitrophenol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'