missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-nitrobenzaldehyde, 97%, Thermo Scientific™

Catalog Number p-7032468
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
157701-72-9
C7H4FNO3
169.111
MFCD00968941
ZFCMKFOFVGHQNE-UHFFFAOYSA-N
2-fluoro-4-nitro-benzaldehyde, benzaldehyde, 2-fluoro-4-nitro, 3-fluoro-4-formylnitrobenzene, fluoronitrobenzaldehyde, pubchem17013, intermediates-zcf02192, 2-floro-4-nitrobenzaldehyde, 4-nitro-2-fluorobenzaldehyde, ksc494i2b, 4-nitro-2-fluoro-benzaldehyde
4562919
2-fluoro-4-nitrobenzaldehyde
C1=CC(=C(C=C1[N+](=O)[O-])F)C=O
This item is not returnable. View return policy
157701-72-9
C7H4FNO3
169.111
MFCD00968941
ZFCMKFOFVGHQNE-UHFFFAOYSA-N
2-fluoro-4-nitro-benzaldehyde, benzaldehyde, 2-fluoro-4-nitro, 3-fluoro-4-formylnitrobenzene, fluoronitrobenzaldehyde, pubchem17013, intermediates-zcf02192, 2-floro-4-nitrobenzaldehyde, 4-nitro-2-fluorobenzaldehyde, ksc494i2b, 4-nitro-2-fluoro-benzaldehyde
4562919
2-fluoro-4-nitrobenzaldehyde
C1=CC(=C(C=C1[N+](=O)[O-])F)C=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 98°C to 100°C
Quantity 250 mg
Sensitivity Air Sensitive
Formula Weight 169.1
Percent Purity 97%
Chemical Name or Material 2-Fluoro-4-nitrobenzaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'