Learn More
2-Fluoro-4-methylaniline, 99%, Thermo Scientific Chemicals
Catalog Number p-7023240
Click to view available options
Quantity:
5 g
25 g
452-80-2
C7H8FN
125.15
MFCD00040975
ZQEXBVHABAJPHJ-UHFFFAOYSA-N
2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
67984
2-fluoro-4-methylaniline
CC1=CC=C(N)C(F)=C1
452-80-2
C7H8FN
125.15
MFCD00040975
ZQEXBVHABAJPHJ-UHFFFAOYSA-N
2-fluoro-4-methyaniline, 2-fluoro-p-toluidine, 4-amino-3-fluorotoluene, p-toluidine, 2-fluoro, benzenamine, 2-fluoro-4-methyl, 2-fluoro-4-methyl aniline, 2-fluoro-4-methyl-aniline, 2-fluoro-4-methyl-phenylamine, 2-fluoro-4-methylphenylamine, benzenamine, 2-fluoro-4-methyl-9ci
67984
2-fluoro-4-methylaniline
CC1=CC=C(N)C(F)=C1
2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Fluoro-4-methylaniline is used in the preparation of 6-chloro-5-fluoroindole via Leimgruber-Batcho reaction. It is also used in the preparation of an (S)-amino alcohol, 2-amino-3-(2-fluoro-4-methylphenyl)-propan-1-ol.
Solubility
Not miscible in water.
Notes
Store in a cool, dry, conditions in well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 1.108
Boiling Point 70°C to 71°C (7 mmHg)
Flash Point 79°C (174°F)
Refractive Index 1.533
Quantity 5 g
UN Number UN2810
Beilstein 2637578
Solubility Information Not miscible in water.
Formula Weight 125.15
Percent Purity 99%
Chemical Name or Material 2-Fluoro-4-methylaniline