missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-4-methoxycarbonylphenylboronic acid, 98%, Thermo Scientific™

Catalog Number p-7077585
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
603122-84-5
C8H8BFO4
197.96
MFCD08436034
BKWRLCIYMAYFPA-UHFFFAOYSA-N
2-fluoro-4-methoxycarbonyl phenylboronic acid, 2-fluoro-4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methoxycarbonyl phenyl boronic acid, o-fluoro p-methoxy carbonyl phenyl boronic acid, pubchem17434, acmc-209mj4, 2-fluoro-4-methoxycarbonyl phenylboronicacid, 2-fluoro-4-methoxycarbonyl-phenylboronic acid
24824345
(2-fluoro-4-methoxycarbonylphenyl)boronic acid
COC(=O)C1=CC(F)=C(C=C1)B(O)O
This item is not returnable. View return policy
603122-84-5
C8H8BFO4
197.96
MFCD08436034
BKWRLCIYMAYFPA-UHFFFAOYSA-N
2-fluoro-4-methoxycarbonyl phenylboronic acid, 2-fluoro-4-methoxycarbonyl benzeneboronic acid, 4-methoxycarbonyl-2-fluorophenylboronic acid, 2-fluoro-4-methoxycarbonyl phenyl boronic acid, o-fluoro p-methoxy carbonyl phenyl boronic acid, pubchem17434, acmc-209mj4, 2-fluoro-4-methoxycarbonyl phenylboronicacid, 2-fluoro-4-methoxycarbonyl-phenylboronic acid
24824345
(2-fluoro-4-methoxycarbonylphenyl)boronic acid
COC(=O)C1=CC(F)=C(C=C1)B(O)O
Melting Point 127.0°C to 129.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 197.96
Percent Purity 98%
Chemical Name or Material 2-Fluoro-4-methoxycarbonylphenylboronic acid
compliance-icons
Product Identifier
  • 2-Fluoro-4-methoxycarbonylphenylboronic acid, 98%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'